ENA238712500 Enamine

4-fluoro-5-(1H-pyrazol-1-yl)benzene-1,2-diamine

  • MDL number MFCD09809686
  • Cas Number : 1052598-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101...103
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50575

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50575
Availability details for
Quantity
  • Availability
  • Add to Cart