ENA238712502 Enamine

3-(bromomethyl)-1-benzothiophene

  • MDL number MFCD00098683
  • Cas Number : 1196-19-6
  • Molecular Weight : 227.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 62-64
Iupac Name 3-(bromomethyl)-1-benzothiophene
Inchi Code 1S/C9H7BrS/c10-5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6H,5H2
Inchi Key WYKHJZSILIVQKU-UHFFFAOYSA-N

Description

Melting Point 62-64
Purity 95
Storage temp. -70
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-50577

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-50577
pictograms GHS05,GHS07
hstatements H302,H314
Availability details for
Quantity
  • Availability
  • Add to Cart