ENA238712503 Enamine

2-(1-bromoethyl)-1,3-difluorobenzene

  • MDL number MFCD11180320
  • Cas Number : 113212-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-50578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50578
Availability details for
Quantity
  • Availability
  • Add to Cart