ENA238712510 Enamine

3',4'-dichloro-[1,1'-biphenyl]-3-amine

  • MDL number MFCD09042296
  • Cas Number : 926201-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50587
Availability details for
Quantity
  • Availability
  • Add to Cart