ENA238712513 Enamine

(1R)-1-(4-propylphenyl)ethan-1-ol

  • MDL number MFCD09863775
  • Cas Number : 105364-41-8
  • Molecular Weight : 164.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50597
Availability details for
Quantity
  • Availability
  • Add to Cart