ENA238712516 Enamine

1-(6-amino-3,4-dihydro-2H-1,4-benzoxazin-4-yl)-2,2-dimethylpropan-1-one

  • MDL number MFCD12912873
  • Cas Number : 1193387-38-0
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2,2-dimethylpropanoyl)-3,4-dihydro-2H-1,4-benzoxazin-6-amine
Inchi Code 1S/C13H18N2O2/c1-13(2,3)12(16)15-6-7-17-11-5-4-9(14)8-10(11)15/h4-5,8H,6-7,14H2,1-3H3
Inchi Key SOXZPKHZFLAKRK-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-50618

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-50618
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart