ENA238712532 Enamine

methyl 4-[(4-methoxypiperidin-1-yl)methyl]benzoate

  • MDL number MFCD12457061
  • Cas Number : 1096353-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50638
Availability details for
Quantity
  • Availability
  • Add to Cart