ENA238712535 Enamine

2-(furan-2-yl)-1H-1,3-benzodiazol-5-amine dihydrochloride

  • MDL number MFCD01828452
  • Cas Number : 471270-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50641
Availability details for
Quantity
  • Availability
  • Add to Cart