ENA238712540 Enamine

(3-methyl-1,2,4-oxadiazol-5-yl)methanamine hydrochloride

  • MDL number MFCD12028451
  • Cas Number : 253196-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197...199
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50657

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-50657
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart