ENA238712548 Enamine

1-[4-(aminomethyl)phenyl]piperidine-3-carboxamide

  • MDL number MFCD11135264
  • Cas Number : 1019625-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50667

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50667
Availability details for
Quantity
  • Availability
  • Add to Cart