ENA238712552 Enamine

6-(cyclopropylamino)pyridine-3-carboxylic acid

  • MDL number MFCD11132008
  • Cas Number : 960060-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50671
Availability details for
Quantity
  • Availability
  • Add to Cart