ENA238712553 Enamine

4-(2-methylpropoxy)piperidine hydrochloride

  • MDL number MFCD08689809
  • Cas Number : 903891-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50672

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50672
Availability details for
Quantity
  • Availability
  • Add to Cart