ENA238712556 Enamine

2-(4-ethyl-5-methyl-1,3-thiazol-2-yl)acetonitrile

  • MDL number MFCD09933769
  • Cas Number : 1016781-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50675
Availability details for
Quantity
  • Availability
  • Add to Cart