ENA238712563 Enamine

4-(pyridin-2-yl)-1,2,5-oxadiazol-3-amine

  • MDL number MFCD08273505
  • Cas Number : 890095-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50851

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50851
Availability details for
Quantity
  • Availability
  • Add to Cart