ENA238712567 Enamine

5-(chloromethyl)-1-[4-(trifluoromethyl)phenyl]-1H-1,2,3,4-tetrazole

  • MDL number MFCD11134247
  • Cas Number : 1094265-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50858
Availability details for
Quantity
  • Availability
  • Add to Cart