ENA238712574 Enamine

2-bromo-4-(chloromethyl)-1-fluorobenzene

  • MDL number MFCD11156114
  • Cas Number : 78239-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50873
Availability details for
Quantity
  • Availability
  • Add to Cart