ENA238712575 Enamine

1-(1,4-diazepan-1-yl)propan-1-one

  • MDL number MFCD09048622
  • Cas Number : 926233-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50874
Availability details for
Quantity
  • Availability
  • Add to Cart