ENA238712583 Enamine

2-[ethyl(phenyl)amino]acetic acid hydrochloride

  • MDL number MFCD12912886
  • Cas Number : 21911-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50890
Availability details for
Quantity
  • Availability
  • Add to Cart