ENA238712589 Enamine

6-bromo-4-chloroquinoline-3-carbonitrile

  • MDL number MFCD09261323
  • Cas Number : 364793-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146...148
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50896

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50896
Availability details for
Quantity
  • Availability
  • Add to Cart