ENA238712600 Enamine

2-(methylamino)-N-(1,3-thiazol-2-yl)acetamide hydrochloride

  • MDL number MFCD12912822
  • Cas Number : 1193388-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50262
Availability details for
Quantity
  • Availability
  • Add to Cart