ENA238712607 Enamine

2-(phenoxymethyl)aniline

  • MDL number MFCD09025893
  • Cas Number : 78584-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50266

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50266
Availability details for
Quantity
  • Availability
  • Add to Cart