ENA238712620 Enamine

3-[(prop-2-yn-1-yl)amino]-1lambda6-thiolane-1,1-dione hydrochloride

  • MDL number MFCD08706425
  • Cas Number : 915402-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50270
Availability details for
Quantity
  • Availability
  • Add to Cart