ENA238712640 Enamine

2-chloro-1-N,1-N-dimethylbenzene-1,4-diamine

  • MDL number MFCD09260796
  • Cas Number : 6085-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72...73
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50275

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50275
Availability details for
Quantity
  • Availability
  • Add to Cart