ENA238712644 Enamine

2-[(3,5-difluorophenyl)formamido]acetic acid

  • MDL number MFCD08445021
  • Cas Number : 923691-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50999
Availability details for
Quantity
  • Availability
  • Add to Cart