ENA238712649 Enamine

4-ethoxy-2-methylaniline

  • MDL number MFCD08687711
  • Cas Number : 114766-05-1
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-50185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50185
Availability details for
Quantity
  • Availability
  • Add to Cart