ENA238712655 Enamine

1-(3,4-dichlorophenyl)-3,3,3-trifluoropropan-1-amine

  • MDL number MFCD11641933
  • Cas Number : 1094525-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51012
Availability details for
Quantity
  • Availability
  • Add to Cart