ENA238712661 Enamine

3-(4-bromo-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD03419655
  • Cas Number : 925146-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112...114
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51019

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-51019
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart