ENA238712666 Enamine

(2-ethoxy-5-nitrophenyl)methanol

  • MDL number MFCD08460626
  • Cas Number : 1039913-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102...104
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51024

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51024
Availability details for
Quantity
  • Availability
  • Add to Cart