ENA238712668 Enamine

3-amino-N-cyclohexylpropanamide

  • MDL number MFCD09726123
  • Cas Number : 115012-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 38...40
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51026

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51026
Availability details for
Quantity
  • Availability
  • Add to Cart