ENA238712674 Enamine

tert-butyl(pyridin-3-ylmethyl)amine

  • MDL number MFCD00716871
  • Cas Number : 97266-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50280

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50280
Availability details for
Quantity
  • Availability
  • Add to Cart