ENA238712680 Enamine

2,4,6-trifluorobenzene-1-thiol

  • MDL number MFCD11189397
  • Cas Number : 130922-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202...212
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51041

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-51041
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart