ENA238712683 Enamine

methyl 4-[4-(morpholin-4-yl)phenyl]-2,4-dioxobutanoate

  • MDL number MFCD12912916
  • Cas Number : 1181537-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51054
Availability details for
Quantity
  • Availability
  • Add to Cart