ENA238712694 Enamine

(1,2-benzoxazol-3-yl)methanesulfonamide

  • MDL number MFCD00865316
  • Cas Number : 68291-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50288

Safety Information

signalword Warning
pcodes P201,P202,P261,P264,P270,P271,P281,P301+P312,P304+P340,P308+P313,P312,P330,P403+P233,P405,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-50288
s_phrases_eu 26-36
pictograms GHS07,GHS08
hstatements H302,H336,H361d
Availability details for
Quantity
  • Availability
  • Add to Cart