ENA238712699 Enamine

3-(3-chloropropyl)imidazolidine-2,4-dione

  • MDL number MFCD09733521
  • Cas Number : 271581-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-51072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51072
Availability details for
Quantity
  • Availability
  • Add to Cart