ENA238712702 Enamine

{[3,5-dimethyl-4-(2,2,2-trifluoroethoxy)phenyl]methyl}(methyl)amine

  • MDL number MFCD11629853
  • Cas Number : 1095218-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51075
Availability details for
Quantity
  • Availability
  • Add to Cart