ENA238712706 Enamine

2-(3-methylbutoxy)ethan-1-ol

  • MDL number MFCD12545833
  • Cas Number : 7521-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-51079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51079
Availability details for
Quantity
  • Availability
  • Add to Cart