ENA238712708 Enamine

2-(4-bromothiophen-2-yl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD11179457
  • Cas Number : 1094456-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51083
Availability details for
Quantity
  • Availability
  • Add to Cart