ENA238712714 Enamine

5-(propan-2-yl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD11155443
  • Cas Number : 6280-28-0
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-100
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-51095

Safety Information

signalword Warning
pcodes P210,P240,P241,P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P370+P378,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-51095
pictograms GHS02,GHS07
hstatements H228,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart