ENA238712722 Enamine

3-methanesulfonamido-2,4,6-trimethylbenzene-1-sulfonyl chloride

  • MDL number MFCD09047952
  • Cas Number : 926272-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51103

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51103
Availability details for
Quantity
  • Availability
  • Add to Cart