ENA238712724 Enamine

2-(1-chloroethyl)-5-methyl-1,3,4-oxadiazole

  • MDL number MFCD09040992
  • Cas Number : 926207-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50188
Availability details for
Quantity
  • Availability
  • Add to Cart