ENA238712728 Enamine

5-methyl-1-(3-methylphenyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD02168479
  • Cas Number : 210159-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51125
Availability details for
Quantity
  • Availability
  • Add to Cart