ENA238712732 Enamine

1-(methylamino)-3-[4-(trifluoromethyl)phenoxy]propan-2-ol

  • MDL number MFCD10693596
  • Cas Number : 1152538-73-2
  • Molecular Weight : 249.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51132
Availability details for
Quantity
  • Availability
  • Add to Cart