ENA238712739 Enamine

1-[(2-methoxyphenyl)methyl]piperidin-4-amine

  • MDL number MFCD09947597
  • Cas Number : 342894-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51139
Availability details for
Quantity
  • Availability
  • Add to Cart