ENA238712742 Enamine

1-[(tert-butoxy)carbonyl]-4-phenoxypiperidine-4-carboxylic acid

  • MDL number MFCD12912928
  • Cas Number : 1193388-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51144
Availability details for
Quantity
  • Availability
  • Add to Cart