ENA238712752 Enamine

N-[4-(1-aminoethyl)phenyl]-4-fluorobenzamide

  • MDL number MFCD09737062
  • Cas Number : 954269-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51167

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51167
Availability details for
Quantity
  • Availability
  • Add to Cart