ENA238712762 Enamine

N-(3-aminophenyl)-5-methyl-1,2-oxazole-4-carboxamide hydrochloride

  • MDL number MFCD12912933
  • Cas Number : 1181867-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51178
Availability details for
Quantity
  • Availability
  • Add to Cart