ENA238712763 Enamine

N-(4-aminophenyl)-5-methyl-1,2-oxazole-4-carboxamide hydrochloride

  • MDL number MFCD12912934
  • Cas Number : 1193388-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51179

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51179
Availability details for
Quantity
  • Availability
  • Add to Cart