ENA238712772 Enamine

3-fluoro-4-(4-methylpiperazin-1-yl)benzaldehyde

  • MDL number MFCD11156351
  • Cas Number : 868770-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51200
Availability details for
Quantity
  • Availability
  • Add to Cart