ENA238712778 Enamine

ethyl 5-sulfamoyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD09737917
  • Cas Number : 96543-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51207
Availability details for
Quantity
  • Availability
  • Add to Cart