ENA238712780 Enamine

4-(methylamino)benzamide hydrochloride

  • MDL number MFCD12912937
  • Cas Number : 1193389-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51209
Availability details for
Quantity
  • Availability
  • Add to Cart